ContactUsLinkComponent
STEMdiff™ Midbrain Neuron Maturation Kit
STEMdiff™ Midbrain Neuron Maturation Kit
Catalog # STMC100-0041
CAS Number:  
undefined
STEMdiff™ Midbrain Neuron Maturation Kit
Catalog # STMC100-0041
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    STEMdiff™ Midbrain neuron maturation kit
Specifications
About this item
Maturation kit for generation of functional midbrain-type neurons from human ES and iPS cell-derived neuronal precursor cells.
The STEMdiff™ Midbrain Neuron Maturation Kit is used in conjunction with the STEMdiff™ Midbrain Neuron Differentiation Kit to generate a mixed population of midbrain-type neurons from neural progenitor cells derived from human pluripotent stem cells. These media will produce a population of midbrain neurons (≥ 15% TH-positive dopaminergic neurons; ≥ 90% class III βtubulin-positive neurons; < 10% GFAP-positive astrocytes). Cells derived using these products are versatile tools for modeling human neurological development and disease, drug screening, toxicity testing, and cell therapy validation.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').