Orders LinkContactUsLinkComponent
StemSpan™ NK Cell Generation Kit
StemSpan™ NK Cell Generation Kit
StemSpan™ NK cell generation kit, 1 KIT
Catalog # STMC09960
Marketsource
StemSpan™ NK Cell Generation Kit
Catalog # STMC09960
Marketsource
CAS Number:  
StemSpan™ NK Cell Generation Kit
StemSpan™ NK Cell Generation Kit
StemSpan™ NK cell generation kit, 1 KIT
CAS Number:  
Price
1 820,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents
For expansion and differentiation of human CD34+ hematopoietic progenitor cells to NK cells.
Differentiate CD34+ cells isolated from cord blood (CB) and bone marrow (BM) to natural killer (NK) cells—without the use of serum or stromal cells—by using StemSpan™ NK Cell Generation Kit. This kit promotes the expansion of thousands of CD56+ NK cells in cultures initiated with CD34+ human CB cells. StemSpan™ Lymphoid Progenitor Expansion Supplement (10X) contains a combination of recombinant human cytokines and other additives formulated to selectively promote the expansion and differentiation of CD34+ cells isolated from human CB and BM samples to lymphoid progenitor cells when used in combination with StemSpan™ SFEM II medium, and plates coated with StemSpan™ Lymphoid Differentiation Coating Material (100X). Subsequently, StemSpan™ NK Cell Differentiation Supplement (100X) enables differentiation of lymphoid progenitor cells to NK cells. For convenience, all the components of the kits are also available individually.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Specifications
Specifications
  • Pk:
    1 KIT
  • Description:
    StemSpan™ NK cell generation kit
Specifications