ContactUsLinkComponent
StemSpan™ T Cell Progenitor Maturation Supplement (10X)
StemSpan™ T Cell Progenitor Maturation Supplement (10X)
Catalog # STMC09930
CAS Number:  
undefined
StemSpan™ T Cell Progenitor Maturation Supplement (10X)
Catalog # STMC09930
CAS Number:  
Specifications
  • Pk:
    12,5 mL
  • Description:
    StemSpan™ T Cell progenitor maturation supplement (10X)
Specifications
About this item
Supplement for the maturation of T cell progenitors to T cells.
Support the maturation of lymphoid progenitor cells to double-positive (DP) T cells using StemSpan™ T Cell Progenitor Maturation Supplement (10X). For convenience, this supplement is available either individually or as part of the StemSpan™ T Cell Generation Kit or the STEMdiff™ T Cell Kit. In this protocol, CD34+ cells are first differentiated to CD7+CD5+ lymphoid progenitor cells, using StemSpan™ Lymphoid Progenitor Expansion Supplement (10X), and then further directed to differentiate to DP T cells.
StemSpan™ T Cell Progenitor Maturation Supplement (10X) is intended for use in combination with StemSpan™ SFEM II medium, and on plates coated with StemSpan™ Lymphoid Differentiation Coating Material (100X). It also supports the maturation of DP T cells to CD8 T cells with additional stimuli. For detailed information on protocols for the expansion and differentiation of CD34+ cells with StemSpan™, please explore the technical manual and educational materials.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').