ContactUsLinkComponent
BrainPhys™ Neuronal Medium N2-A & SM1 Kit
BrainPhys™ Neuronal Medium N2-A & SM1 Kit
Catalog # STMC05793
CAS Number:  
undefined
BrainPhys™ Neuronal Medium N2-A & SM1 Kit
Catalog # STMC05793
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    BrainPhys™ Neuronal medium N2-A & SM1 kit
Specifications
About this item
Kit including BrainPhys™ Neuronal Medium, SM1 Neuronal Supplement, and N2 Supplement-A for culture of ES/iPS cell-derived neurons.
Culture, differentiate, and mature neurons derived from human embryonic stem (ES) or induced pluripotent stem (iPS) cells in a complete medium optimised to promote, rather than inhibit neuronal activity. For your convenience, BrainPhys™ Neuronal Medium N2-A & SM1 Kit includes BrainPhys™ Neuronal Medium (basal medium) and supplements for culturing human ES/iPS-derived neuronal progenitor cells. Based on the formulation by Bardy and Gage (Bardy et al. PNAS, 2015), serum-free BrainPhys™ Neuronal Medium mimics the extracellular environment of the central nervous system (CNS) to yield a higher proportion of synaptically active neurons.
Brewer’s B27-based NeuroCult™ SM1 Neuronal Supplement ensures cell health and encourages neurite outgrowth and branching in short- and long-term serum-free cultures, and N2 Supplement-A supports the in vitro differentiation of ES/iPS-derived cells to neuronal subtypes. For lineage-specific differentiation requiring growth factors, please see BrainPhys™ hPSC Neuron Kit, which also includes BDNF and GDNF. To avoid shocking cells with media changes, also use BrainPhys™ medium when performing functional assays, such as microelectrode array-based recordings or live-fluorescent imaging. View our additional resources to learn more about the BrainPhys™system.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').