ContactUsLinkComponent
NeuroCult™ SM1 Supplement without Antioxidants
NeuroCult™ SM1 Supplement without Antioxidants
Catalog # STMC05732
CAS Number:  
undefined
NeuroCult™ SM1 Supplement without Antioxidants
Catalog # STMC05732
CAS Number:  
Specifications
  • Pk:
    10 mL
  • Description:
    NeuroCult™ SM1 without antioxidants
Specifications
About this item
Serum-free neural supplement without antioxidants.
NeuroCult™ SM1 without Antioxidants is an optimised serum-free culture supplement, based on the published B27 formulation, with antioxidants (tocopherol, tocopherol acetate, superoxide dismutase, catalase, and glutathione) removed. NeuroCult™ SM1 without Antioxidants can be used in protocols where antioxidants may interfere with oxidative stress, or similar assays used in neurodegenerative research. In combination with a basal medium of choice, NeuroCult™ SM1 without Antioxidants can be used for neural progenitor cell expansion and the study of neural stem and progenitor cell differentiation.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').