ContactUsLinkComponent
STEMdiff™ Mesenchymal Progenitor Kit
STEMdiff™ Mesenchymal Progenitor Kit
Catalog # STMC05240
CAS Number:  
undefined
STEMdiff™ Mesenchymal Progenitor Kit
Catalog # STMC05240
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    STEMdiff™ Mesenchymal progenitor kit
Specifications
About this item
Defined culture kit for derivation and expansion of mesenchymal progenitor cells.
STEMdiff™ Mesenchymal Progenitor Kit is a defined culture kit consisting of animal component-free (ACF) induction medium, expansion medium, and attachment substrate. It is optimised for the derivation of cells with mesenchymal progenitor cell (MPC)-like properties from human embryonic stem (ES) cells or induced pluripotent stem (iPS) cells. This kit provides a complete workflow of defined reagents for derivation and expansion of human ES- or iPS-derived MPCs. CollPlant is the manufacturer of the rhCollagen component of the cell attachment substrate. The Product may be used for research purposes only. Please contact STEMCELL for any clinical or commercial application of the Product.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').