ContactUsLinkComponent
MethoCult™ H4535 Enriched without EPO
MethoCult™ H4535 Enriched without EPO
Catalog # STMC04535
CAS Number:  
undefined
MethoCult™ H4535 Enriched without EPO
Catalog # STMC04535
CAS Number:  
Specifications
  • Pk:
    100 mL
  • Description:
    MethoCult™ H4535 enriched without EPO
Specifications
About this item
Pre-aliquoted methylcellulose-based medium with recombinant cytokines (without EPO) for human cells. 24×3 ml vials.
MethoCult™ H4535 Enriched Without EPO (MethoCult™ GF+ H4535) is a complete methylcellulose-based medium for the growth and enumeration of hematopoietic progenitor cells in colony-forming unit (CFU) assays of human bone marrow, mobilized peripheral blood, peripheral blood, and cord blood samples. This medium supports optimal growth of granulocyte-macrophage progenitor cells (CFU-GM, CFU-G and CFU-M), and is recommended for use with CD34+ cells and other purified cell populations.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').