Order Entry
Austria
Orders LinkContactUsLinkComponent
Iscove's Modified Dulbecco's Medium

Iscove's Modified Dulbecco's Medium

Iscove's modified dulbecco's medium, 500 mL
Catalog # STMC36150
Marketsource
Iscove's Modified Dulbecco's Medium
Catalog # STMC36150
Marketsource
CAS Number:  
Iscove's Modified Dulbecco's Medium
Iscove's Modified Dulbecco's Medium

Iscove's modified dulbecco's medium, 500 mL

CAS Number:  
Price
249,00 €
Log in for availability
1 * 500 mL
Product Details & Documents

Iscove's Modified Dulbecco's Medium (IMDM) with 25 mm HEPES.

Iscove's Modified Dulbecco's Medium (IMDM) with 25 mm HEPES is recommended for a wide variety of cell culture applications. Selection of suitable nutrient medium is dependent on the type of cell, conditions of culture, and degree of chemical definition required for the cell culture application.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifications

Specifications

  • Pk:
    500 mL
  • Description:
    Iscove's modified dulbecco's medium

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.