ContactUsLinkComponent
STEMdiff™ Cardiomyocyte Expansion Kit
STEMdiff™ Cardiomyocyte Expansion Kit
Catalog # STMC100-1109
CAS Number:  
undefined
STEMdiff™ Cardiomyocyte Expansion Kit
Catalog # STMC100-1109
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    STEMdiff™ Cardiomyocyte expansion kit
Specifications
About this item
A serum-free media kit for the expansion of early-stage human pluripotent stem cell-derived cardiomyocytes.
Expand early-stage human pluripotent stem cell (hPSC)-derived cardiomyocytes consistently using this serum-free kit. This kit generates a large number of functional and highly pure hPSC-derived cardiomyocytes and is compatible with ventricular or atrial cardiomyocytes generated with STEMdiff™ Ventricular Cardiomyocyte Differentiation Kit or STEMdiff™ Atrial Cardiomyocyte Differentiation Kit, respectively. Compared to the typical method of first expanding large numbers of hPSCs followed by multiple rounds of differentiation to yield high numbers of hPSC-derived cardiomyocytes, the STEMdiff™ Cardiomyocyte Expansion kit allows easy reach of cardiomyocyte populations in the billions by simply expanding early-stage hPSC-derived cardiomyocytes with a single round of cardiomyocyte differentiation.
Using this kit, expanded early-stage hPSC-derived cardiomyocytes retain a stable electrical profile and have cTnT percentages over 90% at passage 5. The expanded hPSC-derived cardiomyocytes are ready for high-throughput drug testing, tissue engineering, and regenerative medicine research. Save time and resources by using this first-to-market kit to efficiently expand cardiomyocytes. Create a complete, optimised workflow with these compatible STEMdiff™ cardiomyocyte products: STEMdiff™ Ventricular Cardiomyocyte Differentiation Kit, STEMdiff™ Atrial Cardiomyocyte Differentiation Kit, STEMdiff™ Cardiomyocyte Dissociation Kit, STEMdiff™ Cardiomyocyte Support Medium, STEMdiff™ Cardiomyocyte Freezing Medium, STEMdiff™ Cardiomyocyte Maintenance Kit.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').