Orders LinkContactUsLinkComponent
STEMdiff™ Endothelial Expansion Medium Kit
STEMdiff™ Endothelial Expansion Medium Kit
STEMdiff™ Endothelial expansion medium kit, 1 KIT
Catalog # STMC08007
Marketsource
STEMdiff™ Endothelial Expansion Medium Kit
Catalog # STMC08007
Marketsource
CAS Number:  
STEMdiff™ Endothelial Expansion Medium Kit
STEMdiff™ Endothelial Expansion Medium Kit
STEMdiff™ Endothelial expansion medium kit, 1 KIT
CAS Number:  
Price
583,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents
Serum- and xeno-free medium for expansion of human pluripotent stem cell-derived endothelial cells.
STEMdiff™ Endothelial Expansion Medium Kit includes the xeno-free endothelial expansion basal medium and supplement, and must be used in conjunction with Animal Component-Free Cell Attachment Substrate. STEMdiff™ Endothelial Expansion Medium is optimised for the expansion of hPSC-derived endothelial cells and for the maintenance of endothelial marker expression. For passaging, Animal Component-Free Cell Dissociation Kit is required. NOTE: Heparin Solution is required for preparation of complete STEMdiff™ Endothelial Expansion Medium. CollPlant is the manufacturer of the rhCollagen component of the cell attachment substrate. The Product may be used for research purposes only.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Specifications
Specifications
  • Pk:
    1 KIT
  • Description:
    STEMdiff™ Endothelial expansion medium kit