ContactUsLinkComponent
MyoCult™-SF Attachment Substrate
MyoCult™-SF Attachment Substrate
Catalog # STMC05983
CAS Number:  
undefined
MyoCult™-SF Attachment Substrate
Catalog # STMC05983
CAS Number:  
Specifications
  • Pk:
    100 µG
  • Description:
    MyoCult™ SF attachment substrate
Specifications
About this item
Serum-free attachment substrate for the derivation and expansion of human skeletal muscle progenitor cells (myoblasts).
Ensure high yields when passaging and culturing primary human skeletal muscle progenitor cells (myoblasts) by using MyoCult™-SF Attachment Substrate, which is optimised to support myoblast attachment. This substrate is recommended for culturing primary myoblasts in MyoCult™-SF Expansion Medium; human pluripotent stem cell (hPSC)-derived myoblasts may be cultured on cultureware coated with Corning® Matrigel® hESC-Qualified Matrix. MyoCult™-SF Attachment Substrate is also available as part of the MyoCult™-SF Expansion Supplement Kit (Human), which includes MyoCult™-SF Expansion 10X Supplement (Human). The supplement is combined with a basal medium to prepare MyoCult™-SF Expansion Medium for the culture of primary human and hPSC-derived myoblasts. Myoblasts cultured using MyoCult™-SF Expansion Medium can be differentiated into multinucleated myotubes using MyoCult™ Differentiation Kit (Human).
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').