ContactUsLinkComponent
RSeT™ Medium (2-Component)
RSeT™ Medium (2-Component)
Catalog # STMC05978
CAS Number:  
undefined
RSeT™ Medium (2-Component)
Catalog # STMC05978
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    RSeT™ Medium (2-component)
Specifications
About this item
Defined medium for naïve-like human pluripotent stem cells.
RSeT™ Medium is a defined cell culture medium used for the reversion of primed human pluripotent stem cells (hPSCs) to a naïve-like state and maintenance of naïve-like hPSCs under feeder-dependent and hypoxic conditions. RSeT™ Medium contains pre-screened quality components and does not contain bFGF or TGFβ. It is compatible with human embryonic stem (ES) cells and human induced pluripotent stem (iPS) cells. hPSCs cultured in RSeT™ Medium exhibit features of a naïve-like state such as tightly packed, domed colonies with refractive edges. Key transcripts associated with naïve-like hPSCs such as KLF2, KLF4, and TFCP2L1 show increased expression in hPSCs cultured in RSeT™ Medium. RSeT™ hPSCs can be converted back to a primed state by culture in mTeSR™1 and can then be differentiated with STEMdiff™ products such as STEMdiff™ Definitive Endoderm Kit, STEMdiff™ SMADi Neural Induction Kit, and STEMdiff™ Mesoderm Induction Medium.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').