ContactUsLinkComponent
STEMdiff™ Trilineage Differentiation Kit
STEMdiff™ Trilineage Differentiation Kit
Catalog # STMC05230
CAS Number:  
undefined
STEMdiff™ Trilineage Differentiation Kit
Catalog # STMC05230
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    STEMdiff™ Trilineage differentiation kit
Specifications
About this item
Functional assay kit to assess pluripotency by directed differentiation of human ES and iPS cells to all three germ layers.
The STEMdiff™ Trilineage Differentiation Kit provides a simple culture assay to functionally validate the ability of new or established human embryonic stem (ES) and induced pluripotent stem (iPS) cell lines to differentiate to the three germ layers: ectoderm, mesoderm, and endoderm. This kit includes specialised, complete media and monolayer-based protocols to perform parallel in vitro directed differentiation experiments for each germ layer, clearly and reproducibly establishing trilineage differentiation potential within one week. STEMdiff™ Trilineage Differentiation Kit is intended to be an endpoint assay and is not optimised for the generation of cells for downstream differentiation or other applications. STEMdiff™ Trilineage Differentiation Kit has been optimised to assess cells maintained in mTeSR™1.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').