ContactUsLinkComponent
MegaCult™-C Medium with Cytokines
MegaCult™-C Medium with Cytokines
Catalog # STMC04901
CAS Number:  
undefined
MegaCult™-C Medium with Cytokines
Catalog # STMC04901
CAS Number:  
Specifications
  • Pk:
    24 Vial
  • Description:
    MegaCult™ C medium with cytokines
Specifications
About this item
Medium with cytokines for culture of human and mouse CFU-Mk. 24×2 ml vials.
Use MegaCult™-C Medium with Cytokines for the culture of colony-forming unit-megakaryocyte progenitor cells (CFU-Mk) in human bone marrow, mobilised peripheral blood, and cord blood samples. Available either individually or as part of the MegaCult™-C Collagen and Medium with Cytokines kit, this medium is suitable for use with CD34+ enriched cells, mononuclear cells, and cells isolated by other purification methods. The medium contains recombinant human (rh) IL-3, IL-6, and thrombopoietin. Collagen Solution, compatible with MegaCult™-C and required for preparation of collagen gels for culturing CFU-Mk and for coating cell culture surfaces, is also available for purchase. Required MegaCult™-C Medium may be used with Collagen Solution, available either individually or as part of the MegaCult™-C Collagen and Medium Without Cytokines kit.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').