ContactUsLinkComponent
ClonaCell™-TCS/G418 Kit
ClonaCell™-TCS/G418 Kit
Catalog # STMC03822
CAS Number:  
undefined
ClonaCell™-TCS/G418 Kit
Catalog # STMC03822
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    ClonaCell™ TCS/G418 kit
Specifications
About this item
Kit containing ClonaCell™-TCS medium and antibiotic G418.
This kit is used for selection and cloning applications where the G418 antibiotic is used as the selective agent. ClonaCell™-TCS Medium is a serum-containing, methylcellulose-based semi-solid medium that can be used for selection and cloning of a variety of suspension-adapted and adherent cell lines, such as BHK-1, CHO-K1, CHO-S, CHO-DG44, and HEK-293. This medium contains pre-selected fetal bovine serum (FBS) and bovine serum albumin (BSA). The G418 antibiotic is suitable for use in cell culture for the selection and maintenance of G418-resistant clones when using ClonaCell™-TCS Medium.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').