ClonaCell™-TCS Medium
Catalog # STMC03814
Supplier: STEMCELL Technologies
CAS Number:
Specifications
- Pk:80 mL
- Description:ClonaCell™ TCS medium
Specifications
About this item
Semi-solid methylcellulose-based medium for selection and cloning of suspension-adapted and adherent cells (serum-containing).
- Individual cells are suspended in viscous medium and form physically separated, discrete colonies that are easily isolated
- Monoclonal cell lines are isolated in less time using fewer resources compared with selection and cloning by limiting dilution
- Diverse clones with a wide range of growth rates and productivities form discrete colonies in the viscous medium
- Rare and high-producing clones can be individually isolated more easily using simultaneous selection and cloning in semi-solid medium compared with selection in bulk liquid cultures
ClonaCell™-TCS Medium is a serum-containing methylcellulose-based semi-solid medium that can be used for selecting and cloning a variety of suspension-adapted cell lines, including CHO-S and hybridomas. It can also be used for semi-solid cloning of some cell lines that grow adherently in the presence of serum, including CHO-K1, BHK-1, and HEK-293. This medium contains pre-selected fetal bovine serum (FBS) and bovine serum albumin (BSA) and supports robust growth of a wide variety of cell types. ClonaCell™-TCS Medium does not contain selection agents.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').