MethoCult™ SF H4636 Medium without EPO
Catalog # STMC100-0945
Supplier: STEMCELL Technologies
CAS Number:
Specifications
- Pk:100 mL
- Description:MethoCult™ SF H4636 without EPO
Specifications
About this item
Serum-free methylcellulose-based medium with recombinant cytokines for human hematopoietic cells.
MethoCult™ SF H4636 without EPO is a complete methylcellulose-based medium for the growth and enumeration of hematopoietic progenitor cells in colony-forming unit (CFU) assays of human bone marrow (BM), mobilised peripheral blood (MPB), peripheral blood (PB), and cord blood (CB) samples. MethoCult™ SF H4636 without EPO is formulated to support the optimal growth of granulocyte-macrophage progenitor cells (CFU-GM, CFU-G and CFU-M). It is suitable for use with CD34+ enriched cells, mononuclear cells, and cells isolated by other purification methods.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').