ContactUsLinkComponent
BrainPhys™ Primary Neuron Kit
BrainPhys™ Primary Neuron Kit
Catalog # STMC05794
CAS Number:  
undefined
BrainPhys™ Primary Neuron Kit
Catalog # STMC05794
CAS Number:  
Specifications
  • Pk:
    500 mL
  • Description:
    BrainPhys™ primary neuron kit
Specifications
About this item
Kit including BrainPhys™ neuronal medium, SM1 meuronal supplement and NeuroCult™ neuronal plating medium for culture of primary-derived neurons.
Plate and culture primary neurons of the central nervous system (CNS) in a complete, serum-free medium optimised to promote, rather than inhibit neuronal activity and maturity. For your convenience, BrainPhys™ neuronal medium and SM1 kit includes both serum-free BrainPhys™ neuronal medium (basal medium) and NeuroCult™ SM1 neuronal supplement and NeuroCult™ neuronal plating medium, to provide a complete culture system for primary-tissue-derived neurons. BrainPhys™ neuronal medium mimics the extracellular environment of the CNS to yield a higher proportion of synaptically active neurons.
NeuroCult™ SM1 neuronal supplement is a Brewer's B27-based supplement that ensures cell health and encourages neurite outgrowth and branching in short- and long-term cultures. To avoid shocking cells with media changes, also use BrainPhys™ medium when performing functional assays, such as microelectrode array-based recordings or live-fluorescent imaging. View our additional resources to learn more about the BrainPhys™ system.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').