ContactUsLinkComponent
NeuroCult™ SM1 without Insulin
NeuroCult™ SM1 without Insulin
Catalog # STMC05733
CAS Number:  
undefined
NeuroCult™ SM1 without Insulin
Catalog # STMC05733
CAS Number:  
Specifications
  • Pk:
    10 mL
  • Description:
    NeuroCult™ SM1 without Insulin
Specifications
About this item
Serum-free neural supplement without insulin.
NeuroCult™ SM1 without insulin is an optimised serum-free culture supplement, based on the published B27 formulation, with insulin removed. NeuroCult™ SM1 without insulin can be used in protocols where the effects of insulin are being studied, or where insulin may interfere with cellular function. In combination with a basal medium of choice, NeuroCult™ SM1 without insulin can be used for neural progenitor cell expansion, the study of neural stem and progenitor cell differentiation, and cardiomyocyte and hepatocyte differentiation protocols.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').