ContactUsLinkComponent
StemSpan™ Myeloid Expansion Supplement II (100X)
StemSpan™ Myeloid Expansion Supplement II (100X)
Catalog # STMC02694
CAS Number:  
undefined
StemSpan™ Myeloid Expansion Supplement II (100X)
Catalog # STMC02694
CAS Number:  
Specifications
  • Pk:
    1 mL
  • Description:
    StemSpan™ myeloid expansion supplement II (100X)
Specifications
About this item
Serum-free culture supplement for expansion and differentiation of human monocytes.
StemSpan™ myeloid expansion supplement II (100X) contains a combination of recombinant human cytokines formulated to selectively promote the expansion and differentiation of human myeloid progenitor cells, generating large numbers of CD14+ monocytes from CD34+ cells isolated from human cord blood (CB) samples.
StemSpan™ myeloid expansion supplement II (100X) is intended for use in combination with any of the following StemSpan™ media:
StemSpan™ SFEM
StemSpan™ SFEM II
StemSpan™-XF
StemSpan™-AOF
When added to serum-free medium, StemSpan™ myeloid expansion supplement II typically promotes the production of hundreds of CD14+ monocytes per input CD34+ cell in 14-day liquid cultures initiated with CD34+ human CB cells. See data tab for more details.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').