Order Entry
Austria
ContactUsLinkComponent
3,3'-Di-n-hexyloxacarbocyanine iodide [DiOC6(3) iodide] 98%
Irritant
3,3'-Di-n-hexyloxacarbocyanine iodide [DiOC6(3) iodide] 98%
Certificates
undefined
Irritant
3,3'-Di-n-hexyloxacarbocyanine iodide [DiOC6(3) iodide] 98%

Specification Test Results

  • Appearance (Colour):
    Red
  • Appearance (Form):
    Powder or Chunks
  • Infrared spectrum:
    Conforms to Structure
  • Wavelength:
    483 - 488 nm
  • Extinction Coefficient, c = 0.01 g/L; Methanol:
    ≥120000
  • Carbon:
    59.3 - 62.4%
  • Nitrogen:
    4.6 - 5.1%
  • Purity (HPLC):
    ≥97.5%
  • Solubility (Turbidity):
    Clear to Slightly Hazy
  • Solubility (Colour), c = 25 mg/ml, CHCl3:
    Orange

Specification Test Results

Frequently Bought Together