Tartrazine, Sigma-Aldrich®
HT3024-120ML
:
- Pk:120 mL
- Seq:2470
- Appearance (Colour):Yellow
- Appearance (Form):Solution
- Appearance (Turbidity):Clear
- Wave Length:424 - 430 nm
- Absorbance:1.750 - 2.700
- Performance:Conforms
- Expiration Date Period:24 months
:
:534.36 g/mol
:M-Clarity™ quality level = MQ500 (more info.)
:Room Temperature
:1934-21-0
:C₁₆H₁₂N₄O₉S₂
:MFCD00148908