Order Entry
Austria
ContactUsLinkComponent
Iron hematoxylin, Sigma-Aldrich®
Flammable
Corrosive
Iron hematoxylin, Sigma-Aldrich®
Catalog # HT1079-1SET
CAS Number:  
Flammable
Corrosive
Iron hematoxylin, Sigma-Aldrich®
Catalog # HT1079-1SET
CAS Number:  

Specifications

  • Pk:
    1 SET
  • Seq:
    0125

Specifications

Specification Test Results

  • Certificate of Analysis:
    Accept
  • Expiration Date Period:
    Earliest component expiration date

Specification Test Results

About this item

 :
 :M-Clarity™ quality level = MQ500 (more info.)
Flash Pt:15.55 °C
 :GHS02
 :GHS05
Storage Temperature:Room Temperature
MDL Number:MFCD00284879