Order Entry
Austria
ContactUsLinkComponent
Ethidium homodimer I 2 mM in DMSO ≥95% (by TLC), Sigma-Aldrich®
Ethidium homodimer I 2 mM in DMSO ≥95% (by TLC), Sigma-Aldrich®
Catalog # E1903-.5ML
CAS Number:  
undefined
Ethidium homodimer I 2 mM in DMSO ≥95% (by TLC), Sigma-Aldrich®
Catalog # E1903-.5ML
CAS Number:  

Specifications

  • Pk:
    0,5 mL
  • Seq:
    2040

Specifications

Specification Test Results

  • Appearance (Colour):
    Red
  • Appearance (Form):
    Liquid
  • E
    Lambda Max at 490-496, in H2O
    Run on Ethidium Homodimer before dissolution in DMSO:
    ≥9000
  • Wavelength
    Lambda Max:
    490 - 496 nm
  • Purity (TLC)
    Run on Ethidium Homodimer before dissolution in DMSO:
    ≥95 %

Specification Test Results

About this item

 :
 :856.75 g/mol
 :M-Clarity™ quality level = MQ100
Storage Temperature:Freezer
CAS Number:61926-22-5
Formula:C₄₆H₅₀Cl₄N₈
MDL Number:MFCD00210948