ContactUsLinkComponent
Memosens Conductivity Sensor for Harsh and Aggressive Media, CLS50D
Memosens Conductivity Sensor for Harsh and Aggressive Media, CLS50D
Catalog # ENDRCLS50D-1JF2/0
Supplier:  Endress+Hauser
New Product
undefined
Memosens Conductivity Sensor for Harsh and Aggressive Media, CLS50D
Catalog # ENDRCLS50D-1JF2/0
Supplier:  Endress+Hauser
New Product
Specifications
  • Connection:
    Thread G³/₄ (process)
  • Description:
    Memosens conductivity sensor CLS50D (CLS50D-AA1D21); standard approval; fixed cable; crimp sleeves; 7 m cable length
  • Material:
    PFA
  • Pk:
    1
Specifications
About this item
Digital toroidal conductivity sensor designed for harsh and aggressive media in the chemical industry.
  • High durability and chemical resistance even under harsh conditions
  • Dirt-repellent materials and large sensor opening avoid soiling
  • Wide temperature range and highly resistant materials make the sensor suitable for virtual any application
  • Maximum process safety thanks to Memosens technology
The Indumax CLS50D is a digital inductive conductivity sensor for standard, hazardous and high-temperature applications. It has high chemical resistance and long durability thanks to robust materials (PFA, PEEK). Moreover, it is insensitive to soiling and performs reliable and repeatable even in harsh and aggressive media. Thanks to Memosens, CLS50D combines maximum process and data integrity with simple operation. It resists corrosion and moisture and facilitates predictive maintenance.
Material: PFA, Chemraz, 1.4571 (sensor/seal/adapter)