ContactUsLinkComponent
Conductivity Sensor for Harsh and Aggressive Media, CLS50
Conductivity Sensor for Harsh and Aggressive Media, CLS50
Artikelnr. ENDRCLS50-1056/0
Supplier:  Endress+Hauser
New Product
undefined
Conductivity Sensor for Harsh and Aggressive Media, CLS50
Artikelnr. ENDRCLS50-1056/0
Supplier:  Endress+Hauser
New Product
Specifications
  • Connection:
    Thread G³/₄ (process)
  • Description:
    Conductivity sensor CLS50 (CLS50-G1B2); ATEX II 1G Ex ia IIC T4/T6 Ga approval; 10 m fixed cable, max. 125 °C
  • Material:
    PEEK
  • Pk:
    1
Specifications
About this item
Toroidal conductivity sensor for harsh and aggressive media in the chemical industry.
  • High durability and chemical resistance even under harsh conditions
  • Dirt-repellent materials and large sensor opening avoid soiling
  • Wide temperature range and highly resistant materials make the sensor suitable for virtual any application
  • Robust design for outstanding long operating life
  • Maximum process safety thanks to Memosens technology
The Indumax CLS50 Memosens conductivity sensor is particularly suitable for use in the chemical and process technology sectors. The six-decade measuring range and the excellent chemical resistance properties of the materials in contact with the medium (PFA or PEEK) make it possible to use this sensor in a wide range of applications, such as: concentration measurement of acids and bases, quality monitoring of chemical products in tanks and pipes, phase separation of product/product mixtures.
Material: PEEK, viton, PEEK (sensor/seal/adapter)