Cyclohexanone-2,4-dinitrophenylhydrazone ≥99%, Sigma-Aldrich®
Catalog # 192457-1G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance (Colour):Yellow to Orange
- Appearance (Form):Powder or Crystals
- Infrared spectrum:Conforms to Structure
- Carbon:51.2 - 52.3%
- Nitrogen:19.9 - 20.3%
- Purity (TLC):≥99%
- Solubility (Turbidity), 25 mg/ml (2.5%), CHCl₃:Clear
- Solubility (Colour):Orange
Specification Test Results
About this item
:
:278.27 g/mol
Melting Pt:159...160 °C (lit.)
Storage Temperature:Room Temperature
EINECS:216-458-1
CAS Number:1589-62-4
Formula:C₁₂H₁₄N₄O₄
MDL Number:MFCD00001658