Order Entry
Austria
ContactUsLinkComponent
 
Gram′s fuchsin solution for microscopy, Sigma-Aldrich®
  87794-2.5L-F
 :  
undefined
Gram′s fuchsin solution for microscopy, Sigma-Aldrich®
  87794-2.5L-F
 :  

 

  • Pk:
    2,5 L
  • Seq:
    5096

 

 

  • Appearance (colour):
    Red to very dark red
  • Appearance (form):
    Liquid
  • Thin layer chromatogr.:
    Corresponds to standard
  • Density D20/4:
    0.997 - 1.001
  • Refractive index N20/D:
    1.333 - 1.335
  • Wavelength (1) (UV):
    545 - 555 nm
  • Extinction (1%/1 cm) (1) (UV):
    Report result
  • Wavelength (2) (UV):
    285 - 295 nm
  • Extinction (1%/1 cm) (2) (UV):
    Report result
  • Solvent (UV):
    Etoh 50%
  • Expiry period:
    36 months

 

 

 :
 :0,999 g/ml at 20 °C
 :M-Clarity™ quality level = MQ100
 :Room Temperature
 :MFCD18071399