Order Entry
Austria
ContactUsLinkComponent
Storage Unit with Six Drawer Trays, PM
Storage Unit with Six Drawer Trays, PM
Catalog # MULC31104
Supplier:  MULTIROIR
undefined
Storage Unit with Six Drawer Trays, PM
Catalog # MULC31104
Supplier:  MULTIROIR

Specifications

  • Description:
    Unit: Grey; Drawers: Assorted
  • Pk:
    1
  • W×D×H ext.:
    273×265×189 mm
  • W×D×H int.:
    246×216×17 mm

Specifications

About this item

Discover the block six drawers model PM, ideal for use in the laboratory.

  • Six drawer trays in mixed colours
  • Made from impact-resistant polystyrene
  • Integrated handle on each tray
  • Drawer trays can be stacked on top of each other

The small 6-drawer storage unit is entirely made of high-impact polystyrene. The drawers are removable and can be used as preparation trays. They can also be safely stacked on top of each other.

Lightweight and durable, this drawer unit is suitable for laboratory use for preparing elements or storing materials.