Order Entry
Austria
ContactUsLinkComponent
Anti-CD38 Mouse Monoclonal Antibody [clone: HIT2] (mFluor™ Green 620)
Anti-CD38 Mouse Monoclonal Antibody [clone: HIT2] (mFluor™ Green 620)
Supplier:  AAT BIOQUEST
Certificates
undefined
Anti-CD38 Mouse Monoclonal Antibody [clone: HIT2] (mFluor™ Green 620)
Supplier:  AAT BIOQUEST

Specifications

  • Catalog No:
  • AATB103800U1
  • AATB103800U0
  • Antigen symbol:
  • CD38
  • CD38
  • Antigen name:
  • CD38 antigen
  • CD38 antigen
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • mFluor™ Green 620
  • mFluor™ Green 620
  • Clone:
  • HIT2
  • HIT2
  • Reactivity:
  • Human
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG1
  • IgG1
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Purification:
  • Affinity chromatography
  • Affinity chromatography
  • Cross adsorption:
  • No
  • No
  • Storage temperature:
  • 2...8 °C with minimized light exposure. Do not freeze.
  • 2...8 °C with minimized light exposure. Do not freeze.
  • Shipping temperature:
  • 20
  • 20

Specifications

Frequently Bought Together