Order Entry
Austria
ContactUsLinkComponent
Anti-CD161 Mouse Monoclonal Antibody [clone: HP-3G10] (APC (Allophycocyanin)/iFluor® 750)
Anti-CD161 Mouse Monoclonal Antibody [clone: HP-3G10] (APC (Allophycocyanin)/iFluor® 750)
Supplier:  AAT BIOQUEST
Certificates
undefined
Anti-CD161 Mouse Monoclonal Antibody [clone: HP-3G10] (APC (Allophycocyanin)/iFluor® 750)
Supplier:  AAT BIOQUEST

Specifications

  • Catalog No:
  • AATB116101G0
  • AATB116101G2
  • AATB116101G1
  • Antigen symbol:
  • CD161
  • CD161
  • CD161
  • Antigen name:
  • CD161 antigen
  • CD161 antigen
  • CD161 antigen
  • Antibody type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)/iFluor® 750
  • APC (Allophycocyanin)/iFluor® 750
  • APC (Allophycocyanin)/iFluor® 750
  • Clone:
  • HP-3G10
  • HP-3G10
  • HP-3G10
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Mouse
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Yes
  • Purification:
  • Affinity chromatography
  • Affinity chromatography
  • Affinity chromatography
  • Cross adsorption:
  • No
  • No
  • No
  • Storage temperature:
  • 2...8 °C with minimized light exposure. Do not freeze.
  • 2...8 °C with minimized light exposure. Do not freeze.
  • 2...8 °C with minimized light exposure. Do not freeze.
  • Shipping temperature:
  • 20
  • 20
  • 20

Specifications

Frequently Bought Together