Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4] (iFluor® A7)
Supplier: AAT BIOQUEST
Certificates
About this item
RPA-T4 is an anti-human monoclonal antibody that targets the CD4 antigen.
- Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Caution: For research use only.
Type: Primary
Antigen: CD4 antigen
Clonality: Monoclonal
Clone: RPA-T4
Conjugation: iFluor® A7
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
Specifications
- Catalog No:
- AATB100410S0
- AATB100410S1
- Antigen symbol:
- CD4
- CD4
- Antigen name:
- CD4 antigen
- CD4 antigen
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- iFluor® A7
- iFluor® A7
- Clone:
- RPA-T4
- RPA-T4
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- ImmunoFluorescence:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Purification:
- Affinity chromatography
- Affinity chromatography
- Cross adsorption:
- No
- No
- Storage temperature:
- 2...8 °C with minimised light exposure. Do not freeze.
- 2...8 °C with minimised light exposure. Do not freeze.
- Shipping temperature:
- 20
- 20
Specifications
Frequently Bought Together