Anti-CD4 Mouse Monoclonal Antibody [clone: SK3] (iFluor® 700)
Catalog # AATB100420J0
Supplier: AAT BIOQUEST
Specifications
- Antibody type:Primary
- Antigen name:CD4 antigen
- Antigen symbol:CD4
- Clonality:Monoclonal
- Clone:SK3
- Conjugation:iFluor® 700
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:lgG1 kappa
- Reactivity:Human
- Cross adsorption:No
- Storage temperature:Refrigerated (2...8 °C); Minimise light exposure
- Shipping temperature:20
- Purification:Affinity chromatography
- Pack type:Vial
- Pk:100 Tests
Specifications
About this item
SK3 is an anti-human monoclonal antibody that recognises the CD4 antigen.
- Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Caution: For research use only.
Type: Primary
Antigen: CD4 antigen
Clonality: Monoclonal
Clone: SK3
Conjugation: iFluor® 700
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human