Order Entry
Austria
ContactUsLinkComponent
 
VWR® AVS TITRINORM Buffer Solution, pH 9 (at 20 °C), Borate free
  32037.297
 :  
VWR® AVS TITRINORM Buffer Solution, pH 9 (at 20 °C), Borate free
  32037.297
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Colour:
    Colourless
  • Description:
    Buffer solution pH 9.00 (at 20 °C), borate free
  • pH accuracy:
    ±0,02

 

 

Solution prepared from AnalaR® NORMAPUR® grade analytical reagents.

  • Borate free
  • Reference temperature 20 °C
  • True analytical value on the label to an accuracy of ±0,02 pH units
  • Traceable to SRM from NIST
  • Batch number and packing date for traceability
  • Certificate of Analysis available at website
 : If significantly large volumes are required please contact your local Avantor sales office for a customised offer.