Anti-CD16a Mouse Monoclonal Antibody [clone: 3G8] (PacBlue)
Supplier: AAT BIOQUEST
Certificates
About this item
The 3G8 monoclonal antibody recognises human CD16a, a 50 to 65 kD transmembrane protein typically found on the surface of macrophages, natural killer cells, dendritic cells, monocytes and neutrophils.
- Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Caution: For research use only.
Type: Primary
Antigen: CD16a
Clonality: Monoclonal
Clone: 3G8
Conjugation: PacBlue
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Specifications
- Catalog No:
- AATB101611J1
- AATB101611J0
- Antigen symbol:
- CD16a
- CD16a
- Antigen name:
- CD16a antigen
- CD16a antigen
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PB (Pacific Blue)
- PB (Pacific Blue)
- Clone:
- 3G8
- 3G8
- Reactivity:
- Human
- Host:
- Mouse
- Mouse
- ImmunoFluorescence:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Purification:
- Affinity chromatography
- Affinity chromatography
- Cross adsorption:
- No
- No
- Storage temperature:
- 2...8 °C with minimised light exposure, do not freeze
- 2...8 °C with minimised light exposure, do not freeze
- Shipping temperature:
- 20
- 20
Specifications
Frequently Bought Together