Order Entry
Austria
ContactUsLinkComponent
Anti-CD4 Mouse Monoclonal Antibody [clone: SK3] (mFluor™ Violet 510)
Anti-CD4 Mouse Monoclonal Antibody [clone: SK3] (mFluor™ Violet 510)
Catalog # AATB10042110
Supplier:  AAT BIOQUEST
undefined
Anti-CD4 Mouse Monoclonal Antibody [clone: SK3] (mFluor™ Violet 510)
Catalog # AATB10042110
Supplier:  AAT BIOQUEST

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD4 antigen
  • Antigen symbol:
    CD4
  • Clonality:
    Monoclonal
  • Clone:
    SK3
  • Conjugation:
    mFluor™ Violet 510
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    lgG1 kappa
  • Cross adsorption:
    No
  • Storage temperature:
    2...8 °C with minimised light exposure, do not freeze
  • Shipping temperature:
    20
  • Purification:
    Affinity chromatography
  • Pack type:
    Vial
  • Pk:
    100 Tests

Specifications

About this item

SK3 is an anti-human monoclonal antibody that recognises the CD4 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Caution: For research use only.

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: SK3
Conjugation: mFluor™ Violet 510
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human