Order Entry
Austria
ContactUsLinkComponent
Anti-CD200 Mouse Monoclonal Antibody [clone: OX-104] (mFluor™ Violet 510)
Anti-CD200 Mouse Monoclonal Antibody [clone: OX-104] (mFluor™ Violet 510)
  AATB12000110
undefined
Anti-CD200 Mouse Monoclonal Antibody [clone: OX-104] (mFluor™ Violet 510)
  AATB12000110

 

  • Antibody type:
    Primary
  • Antigen name:
    CD200 antigen
  • Antigen symbol:
    CD200
  • Clonality:
    Monoclonal
  • Clone:
    OX-104
  • Conjugation:
    mFluor™ Violet 510
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    lgG1 kappa
  • Cross adsorption:
    No
  • Storage temperature:
    2...8 °C with minimised light exposure, do not freeze
  • Shipping temperature:
    20
  • Purification:
    Affinity chromatography
  • Pack type:
    Vial
  • Pk:
    100 Tests

 

 

OX-104 is an anti-human monoclonal antibody that forms an immune complex with the CD200 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

 : For research use only.

Type: Primary
Antigen: CD200
Clonality: Monoclonal
Clone: OX-104
Conjugation: mFluor™ Violet 510
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human