Orders LinkContactUsLinkComponent
Assistent® Brushes for Bottles
Assistent® Brushes for Bottles
Assistent® Brushes for Bottles
Certificates
Assistent® Brushes for Bottles
Assistent® Brushes for Bottles
6 Options Available Below
Product Details & Documents
An assortment of brushes, available with nylon or natural bristles, designed for use with laboratory bottles and flasks.
  • For steep-bottom, round-shoulder, and threaded bottles and flasks
Brushes feature a head bundle for better cleaning of the bottoms of bottles and flasks.
Product Family Options
Product Information
  • Bristle material
    Brush head Ø
    Handle material
    Total length
    Availability
    Price
  • Nylon
    30 mm
    Steel wire
    270 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 37,00 €
    37,00 €
    1 * 10 ST
       37,00 €
  • Nylon
    45 mm
    Steel wire
    390 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 43,00 €
    43,00 €
    1 * 10 ST
       43,00 €
  • Nylon
    55 mm
    Steel wire
    420 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 4,70 €
    4,70 €
    1 * 1 ST
       4,70 €
  • Nylon
    60 mm
    Steel wire
    470 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 6,20 €
    6,20 €
    1 * 1 ST
       6,20 €
  • Natural bristle
    15 mm
    Steel wire
    280 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 32,80 €
    32,80 €
    1 * 10 ST
       32,80 €
  • Natural bristle
    90 mm
    Wood
    630 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 9,40 €
    9,40 €
    1 * 1 ST
       9,40 €