Anti-PPP1R18 Rabbit Polyclonal Antibody
Catalog # PRSI64-226
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Phostensin
- Antigen symbol:PPP1R18
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:Amino Acid Range = 192-220
- Form:Liquid
- Gene ID:170954
- Modification:None
- Molecular weight:68 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,44 mg/ml
- Shipping temperature:4 °C
- Immunogen:This PHTNS antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 192-220 amino acids from the N-terminal region of human PHTNS.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Size:400 μl
- Pk:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:50~100. For FACS starting dilution is: 1:10~50.
Type: Primary
Antigen: PPP1R18
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 192-220
Host: Rabbit
Isotype: Ig
Reactivity: Human