Folding Backrest Trolley
Folding backrest trolley, self-assembly, 100 mm Ø wheels, 300 kg, 610×415×900 mm
MULC65021
Trolley with fold-down backrest to help transport bins, boxes and cartons.
- Tray in reinforced PP, 35 mm thick
- 100 mm dia. silent castors
- Two fixed castors + two swivel castors with brakes
- 300 kg load capacity
- Black
- Ideal for 600×400 Europe containers
- Delivered unassembled for easy assembly
This folding backrest trolley is made of polypropylene. It allows for the movement of a load not exceeding 300 kg. It is equipped with four wheels (two fixed and two multidirectional). This trolley is delivered unassembled. A few minutes are required for assembly.
- Capacity:300 kg
- Description:Folding backrest trolley, self-assembly, 100 mm Ø wheels
- W×D×H:610×415×900 mm



