Order Entry
Austria
ContactUsLinkComponent
Anti-PRKAB1 Rabbit Polyclonal Antibody
Catalog # PRSI23-280
Supplier:  ProSci Inc.
Anti-PRKAB1 Rabbit Polyclonal Antibody
Catalog # PRSI23-280
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Protein kinase, AMP-activated, beta 1 non-catalytic subunit
  • Antigen symbol:
    PRKAB1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    5564
  • Antigen synonyms:
    AMPK subunit beta-1|AMPK|HAMPKb|AMPK beta -1 chain|PRKAB1|AMPK beta 1|AMPKb
  • Storage buffer:
    PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
  • Storage temperature:
    Store at −20...+730 °C. Avoid freeze/thaw cycles.
  • Concentration:
    Batch dependent
  • Shipping temperature:
    20
  • Immunogen:
    A synthetic peptide of human PRKAB1
  • Purification:
    Affinity purification
  • Pack type:
    Vial
  • Pk:
    50 µl

Specifications

About this item

Type: Primary
Antigen: PRKAB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat