Anti-TLR3 Mouse Monoclonal Antibody [clone: TLR3.7] (PE (Phycoerythrin))
PRSI76-956
New Product
- Antibody type:Primary
- Antigen name:toll-like receptor 3
- Antigen symbol:TLR3
- Clonality:Monoclonal
- Clone:TLR3.7
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Cross adsorption:No
- Form:liquid
- Gene ID:O15455
- Antigen synonyms:TLR3
- Storage buffer:Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
- Storage temperature:The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:4 °C
- Purification:The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
- Pk:0,025 mg
The TLR3.7 monoclonal antibody specifically reacts with human CD283 (TLR3), a type I transmembrane signaling receptor containing IL-1 receptor like intracellular domain and leucine-rich repeats (LRR) in the extracellular domain. It is expressed by dendritic cells and recognize double-stranded RNA and polyinosine-polycytidylic acid. Upon ligand binding, CD283 induces type I interferon production and activation of the nuclear factor kappa-light-chain-enhancer of activated B cells complex (NF-kappaB).
Type: Primary
Antigen: TLR3
Clonality: Monoclonal
Clone: TLR3.7
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human