Anti-PDCD1 Mouse Monoclonal Antibody [clone: MIH4] (FITC (Fluorescein Isothiocyanate))
PRSI76-973
New Product
- Antibody type:Primary
- Antigen name:programmed cell death 1
- Antigen symbol:PDCD1
- Clonality:Monoclonal
- Clone:MIH4
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Cross adsorption:No
- Form:liquid
- Gene ID:Q15116
- Antigen synonyms:PDCD1
- Storage buffer:Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
- Storage temperature:The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:4 °C
- Purification:The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
- Pk:100 Tests
The MIH4 monoclonal antibody specifically reacts with human Programmed death-1 (PD-1 or CD279), a 50 to 55 kDa glycoprotein. It is expressed on mainly on activated B, T, and myeloid cells. Within the cytoplasmic region, PD-1 contains an Immunoreceptor tyrosine-based inhibitory motif (ITIM) and seems to regulate peripheral tolerance. The lack or mutation of CD279 is linked to autoimmune disorders.
Type: Primary
Antigen: PDCD1
Clonality: Monoclonal
Clone: MIH4
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human