Anti-ICOS Mouse Monoclonal Antibody [clone: ISA-3] (PE (Phycoerythrin)-Cy7®)
Catalog # PRSI76-969
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:inducible T-cell co-stimulator
- Antigen symbol:ICOS
- Clonality:Monoclonal
- Clone:ISA-3
- Conjugation:PE (Phycoerythrin)-Cy7®
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Cross adsorption:No
- Form:liquid
- Gene ID:Q9Y6W8
- Antigen synonyms:ICOS
- Storage buffer:Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
- Storage temperature:The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:4 °C
- Purification:The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
- Pk:100 Tests
Specifications
About this item
The ISA-3 monoclonal antibody specifically reacts with human CD278 or Inducible COStimulatory molecule (ICOS), a 47 to 60 kDa homodimeric glycoprotein also known as H4, AILIM, and CRP-1. It is expressed on activated T cells and a group of thymocytes and is involved in the proliferation, activation, and cytokine production of T cells. When plate-bound, the ISA-3 antibody induces the production of IL-4,5,10 in T cells.
Type: Primary
Antigen: ICOS
Clonality: Monoclonal
Clone: ISA-3
Conjugation: PE (Phycoerythrin)-Cy7®
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human