Order Entry
Austria
ContactUsLinkComponent
Anti-B3GAT1 Mouse Monoclonal Antibody [clone: TBO1] (FITC (Fluorescein Isothiocyanate))
Anti-B3GAT1 Mouse Monoclonal Antibody [clone: TBO1] (FITC (Fluorescein Isothiocyanate))
  PRSI76-636
New Product
Anti-B3GAT1 Mouse Monoclonal Antibody [clone: TBO1] (FITC (Fluorescein Isothiocyanate))
  PRSI76-636
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    beta-1,3-glucuronyltransferase 1
  • Antigen symbol:
    B3GAT1
  • Clonality:
    Monoclonal
  • Clone:
    TBO1
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgM
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    Q9P2W7
  • Antigen synonyms:
    B3GAT1
  • Storage buffer:
    Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
  • Storage temperature:
    The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
  • Shipping temperature:
    4 °C
  • Purification:
    The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
  • Pk:
    25 Tests

 

 

The TBO1 monoclonal antibody specifically binds to human CD57, a 110 kDa glycoprotein expressed on a subset of natural killer lymphocytes, cells, neural cells and striated muscle. It is reported that the molecule is involved in cell-matrix interactions and is upregulated in some disease states.

Type: Primary
Antigen: B3GAT1
Clonality: Monoclonal
Clone: TBO1
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human