Order Entry
Austria
ContactUsLinkComponent
Anti-Il17a Rat Monoclonal Antibody [clone: 17B7] (FITC (Fluorescein Isothiocyanate))
Anti-Il17a Rat Monoclonal Antibody [clone: 17B7] (FITC (Fluorescein Isothiocyanate))
Catalog # PRSI77-006
Supplier:  ProSci Inc.
New Product
Anti-Il17a Rat Monoclonal Antibody [clone: 17B7] (FITC (Fluorescein Isothiocyanate))
Catalog # PRSI77-006
Supplier:  ProSci Inc.
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    interleukin 17A
  • Antigen symbol:
    Il17a
  • Clonality:
    Monoclonal
  • Clone:
    17B7
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • Isotype:
    IgG2a kappa
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    Q62386; Q61453
  • Antigen synonyms:
    Il17a
  • Storage buffer:
    Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
  • Storage temperature:
    The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
  • Shipping temperature:
    4 °C
  • Purification:
    The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
  • Pk:
    0,05 mg

Specifications

About this item

The 17B7 monoclonal antibody specifically binds to mouse and rat IL-17A, a pro-inflammatory cytokine. It is produced by T helper 17 (Th17) cells, a unique subset of IL-23 dependent CD4+ T cells. Interleukin-17A is highly expressed in transplant rejection, asthma, psoriasis, and multiple sclerosis, and enhances the expression of ICAM-1 in human fibroblasts. The homodimer is expressed by activated peripheral CD4+ T lymphocytes. The Interleukin-17A binds to the IL-17 receptors (IL-17R) expressed by mast cells, monocytes and macrophages, fibroblasts, and endothelial and epithelial cells.

Type: Primary
Antigen: Il17a
Clonality: Monoclonal
Clone: 17B7
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse;Rat