Order Entry
Austria
ContactUsLinkComponent
Anti-Ifng Rat Monoclonal Antibody [clone: XMG1.2] (FITC (Fluorescein Isothiocyanate))
Anti-Ifng Rat Monoclonal Antibody [clone: XMG1.2] (FITC (Fluorescein Isothiocyanate))
Catalog # PRSI77-046
Supplier:  ProSci Inc.
New Product
Anti-Ifng Rat Monoclonal Antibody [clone: XMG1.2] (FITC (Fluorescein Isothiocyanate))
Catalog # PRSI77-046
Supplier:  ProSci Inc.
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    interferon gamma
  • Antigen symbol:
    Ifng
  • Clonality:
    Monoclonal
  • Clone:
    XMG1.2
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • Isotype:
    IgG1 kappa
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    P01580
  • Antigen synonyms:
    Ifng
  • Storage buffer:
    Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
  • Storage temperature:
    The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
  • Shipping temperature:
    4 °C
  • Purification:
    The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
  • Pk:
    0,025 mg

Specifications

About this item

The XMG1.2 is a neutralizing antibody that binds with the mouse Interferon-gamma (IFN-gamma) protein, a 15 to 17 kDa cytokine with significant antibacterial, antiviral, and antitumoral properties. When secreted by natural killer cells and by natural killer T lymphocytes, it regulates the immune response and supports adaptive immunity when produced by Th1 or CD8+ T lymphocytes. IFN-gamma plays an important role in the activation, the growth, and the differentiation of the macrophages, B and T lymphocytes, and natural killer cells. It interacts synergically with other cytokines, such as TNF-alpha, to inhibit proliferation of normal and transformed cells. IFN- gamma is the primary cytokine that defines Th-1 cells.The biological activity of IFN-gamma is not affected by glycosylation.

Type: Primary
Antigen: Ifng
Clonality: Monoclonal
Clone: XMG1.2
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG1 kappa
Reactivity: Mouse