Anti-Cd8a Rat Monoclonal Antibody [clone: 2.43] (APC (Allophycocyanin))
PRSI76-657
New Product
- Antibody type:Primary
- Antigen name:CD8 antigen, alpha chain
- Antigen symbol:Cd8a
- Clonality:Monoclonal
- Clone:2.43
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG2b
- Reactivity:Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:P01731
- Antigen synonyms:Cd8a
- Storage buffer:Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
- Storage temperature:The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:4 °C
- Purification:The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
- Pk:0,1 mg
The 2.43 monoclonal antibody specifically reacts with mouse CD8 antigen. CD8a (the alpha chains) form heterodimers with CD8b (the beta chains) or homodimers (alpha-alpha), which occur as receptors on the surface of the majority of thymocytes. A subpopulation of mature T lymphocytes expresses the CD8 alpha beta (alpha beta TCR T cells), and a subpopulation of intestinal intraepithelial lymphocytes and dendritic cells express CD8a without CD8b. CD8 interacts with the mouse major histocompatibility complex class I (MHC class I) molecules on antigen-presenting cells or epithelial cells.
Type: Primary
Antigen: Cd8a
Clonality: Monoclonal
Clone: 2.43
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse