Anti-CD38 Mouse Monoclonal Antibody [clone: HIT2] (VioletFluor® 450)
Catalog # PRSI76-748
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:CD38 molecule
- Antigen symbol:CD38
- Clonality:Monoclonal
- Clone:HIT2
- Conjugation:VioletFluor® 450
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Cross adsorption:No
- Form:liquid
- Gene ID:P28907
- Antigen synonyms:CD38
- Storage buffer:Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
- Storage temperature:The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:4 °C
- Purification:The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
- Pk:100 Tests
Specifications
About this item
The HIT2 monoclonal antibody specifically bind to human CD38, a 45 kDa type II transmembrane glycoprotein. CD38 is expressed on activated T cells, plasma cells, monocytes, macrophages, dendritic cells, and thymocytes. CD38 displays cyclase and hydrolase enzymatic activity and is involved in lymphocyte adhesion, activation, and cell signaling. Its ligand is CD31.
Type: Primary
Antigen: CD38
Clonality: Monoclonal
Clone: HIT2
Conjugation: VioletFluor® 450
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human