Order Entry
Austria
ContactUsLinkComponent
Anti-Pdcd1 Hamster Monoclonal Antibody [clone: J43.1] (PE (Phycoerythrin))
Anti-Pdcd1 Hamster Monoclonal Antibody [clone: J43.1] (PE (Phycoerythrin))
Catalog # PRSI76-978
Supplier:  ProSci Inc.
New Product
Anti-Pdcd1 Hamster Monoclonal Antibody [clone: J43.1] (PE (Phycoerythrin))
Catalog # PRSI76-978
Supplier:  ProSci Inc.
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    programmed cell death 1
  • Antigen symbol:
    Pdcd1
  • Clonality:
    Monoclonal
  • Clone:
    J43.1
  • Conjugation:
    PE (Phycoerythrin)
  • Flow cytometry:
    Yes
  • Host:
    Hamster
  • Isotype:
    IgG
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    Q02242
  • Antigen synonyms:
    Pdcd1
  • Storage buffer:
    Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
  • Storage temperature:
    The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
  • Shipping temperature:
    4 °C
  • Purification:
    The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
  • Pk:
    0,025 mg

Specifications

About this item

The J43.1 monoclonal antibody specifically reacts with mouse CD279, also known as PD-1 (programmed death-1), a 50 to 55 kDa glycoprotein of the Ig superfamily. The PD-1 ligands, PD-L1 (B7-H1) and PD-L2 (B7-H2) are members of the B7 family. Pd-1 contains an Immunoreceptor Tyrosine-based Inhibitory Motif (ITIM) and influences the peripheral tolerances and autoimmune diseases in mice. PD-1 is transiently expressed on CD4/CD8 thymocytes, it is upregulated in apoptotic cells, and it is expressed by activated myeloid and T and B cells. The binding of PD-1 to its ligands is blocked by the J43 antibody, which also enhances contact hypersensitivity and exacerbates acute Graft-versus-host disease, Experimental autoimmune encephalomyelitis and NOD diabetes. PD-1 seems to downregulate the immune response, as the development of splenomegaly and breakdown of peripheral tolerance in PD-1 deficient mice suggests.

Type: Primary
Antigen: Pdcd1
Clonality: Monoclonal
Clone: J43.1
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse